Learn More
Thermo Scientific™ Thermolyne™ Premium Large Muffle Furnaces Accessories
Product Code. 15387668 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Shelf
Shelf Pegs
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15387668 Shelf Pegs Each
11559088 Shelf Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15387668 Supplier Thermo Scientific™ Supplier No. JSX16
  • ONLINE EXCLUSIVE PRICE
    16.40 EUR
    15.50 EUR / Pack of 1
    Save 0.90 EUR
    5% Off
 Restricted Item
Estimated Shipment: 26-02-2027
Use these accessories with Thermolyne Premium Large Muffle Furnaces.
Use Thermo Scientific™ Thermolyne Premium Large Muffle Furnace Accessories to increase the utility of the Large Muffle Furnace.
TRUSTED_SUSTAINABILITY
Specifications
Description Shelf Pegs
For Use With (Equipment) Thermolyne Premium Large Muffle Furnace
Temperature (Metric) 100°C to 1200°C
Type Shelf Pegs
Voltage 0/1 V