Learn More
Epredia™ Ultra Disposable Microtome Blades
Epredia™ ultra microtome blades offer high consistency, impressive durability, dependability, and precise performance.
Brand: Epredia™ 3153735
63.92 EUR valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Additional Details : Weight : 0.15300kg
Description
- High- and low-profile universal blades can be used with any microtome or cryostat on the market
- Unique angle profile that facilitates faster ribboning
- Increased Longevity: Produces an edge that is twice as strong as traditional blades
- Reliable Consistency: New proprietary process ensures a durable and consistent coating
Specifications
High | |
Microtomes, Cryostats | |
Yes | |
34° |