Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Tube Adapters for Four-Place Swing Bucket Bioshield Rotor
Product Code. 10260262 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
20 x 7 mL Tubes
4 x 45/50 mL Tubes
4 x 50 mL Conical Tubes
56 x 1.5/2.0 mL Tubes
Unit Size:
2 sets
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
10260262 4 x 50 mL Conical Tubes Each
10763073 56 x 1.5/2.0 mL Tubes 2 sets
10266033 20 x 7 mL Tubes Each
10215703 4 x 45/50 mL Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10260262 Supplier Thermo Scientific™ Supplier No. 75006461
  • ONLINE EXCLUSIVE PRICE
    151.00 EUR
    140.00 EUR / Pack of 1
    Save 11.00 EUR
    7% Off
In Stock
For use with four-place Bioshield rotor 75-006-435 and with four-place rotor 75-006-445 when equipped with rectangular bucket 75-006-446
Adapters sold individually; order correct quantity to equip rotor.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) LH-4000W and LH-4000 rotor with DoubleSpin bucket
Capacity 4 x 50 mL Conical Tubes
Product Type Tube Adapter
Color Light Green