Learn More
Epredia™ Shandon™ Sequenza™ Immunostaining Center Slide Rack
Product Code. 10098889 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Slide Rack
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10098889 Slide Rack Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10098889 Supplier Epredia™ Supplier No. 73310017
  • ONLINE EXCLUSIVE PRICE
    862.00 EUR
    270.23 EUR / Pack of 1
    Save 591.77 EUR
    68% Off
In Stock
Slide Rack, for use with the Shandon™ Sequenza™ Immunostaining Center.
  • Slide Rack holds 10 Shandon Coverplate™ assemblies, which ensure consistency and excellent staining quality
TRUSTED_SUSTAINABILITY
Specifications
Product Type Slide Rack
Description Cassette with 10 staining positions
For Use With (Equipment) Shandon™ Sequenza™ Immunostaining Center