Learn More
Thermo Scientific™ Round Buckets for 4-Place Swinging Bucket Rotor
Product Code. 10210362 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Swinging bucket rotor
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10210362 Swinging bucket rotor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10210362 Supplier Thermo Scientific™ Supplier No. 75007555
  • ONLINE EXCLUSIVE PRICE
    325.00 EUR
    297.00 EUR / Pack of 1
    Save 28.00 EUR
    8% Off
In Stock
4 x 100mL capacity buckets fit the Thermo Scientific Swinging 4-place rotor (75-007-591)
Order Info
Sold individually; order correct quantity to equip rotor. Biocontainment lids (75-007-598) sold separately. Optional adapters accept assorted tube styles and volumes from 1.5 to 50mL. No adapter is required for 100mL bottle.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Swinging bucket rotor