Learn More
Thermo Scientific™ Remel™ Brevundimonas diminuta ATCC™ 11568, Culti-Loops™
Description
The loop may be dissolved in liquid medium, Culti-Loop Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents, and identification kits, and for the evaluation of bacteriological procedures. MycoBacti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms.
Order Info
Culti-Loop products are individually sealed in a foil pouch and packaged five loops per package. Each Mycobacti-Loop is individually sealed in a foil pouch and packaged one loop per package. BactiDisk products are packaged 10 disks per vial.
Specifications
Specifications
| ATCC® | 11568™* |
| Product Type | Culti-Loop |
| Strain Designations | DSM 7234 [CCEB 513, CCM 2657, CCTM 1850, CCUG 1427, CECT 317, CIP 63.27, HNCMB 173005, IAM 1513, IMET 10409, JCM 2788, LMG 2089, NCIB 9393, NCTC 8545, NRRL B-1496, R. Hugh 342, USCC 1337, VKM B-893] |
| Species | Brevundimonas diminuta |
Culti-Loops are CE marked.