Learn More
Thermo Scientific™ Remel™ Brevundimonas diminuta ATCC™ 11568, Culti-Loops™
Product Code. 12965833
Change view
Click to view available options
Brevundimonas diminuta ATCC 11568
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12965833 Brevundimonas diminuta ATCC 11568
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12965833 Supplier Thermo Scientific™ Supplier No. R4607219
The loop may be dissolved in liquid medium, Culti-Loop Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents, and identification kits, and for the evaluation of bacteriological procedures. MycoBacti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms.
Order Info
Culti-Loop products are individually sealed in a foil pouch and packaged five loops per package. Each Mycobacti-Loop is individually sealed in a foil pouch and packaged one loop per package. BactiDisk products are packaged 10 disks per vial.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 11568™*
Product Type Culti-Loop
Strain Designations DSM 7234 [CCEB 513, CCM 2657, CCTM 1850, CCUG 1427, CECT 317, CIP 63.27, HNCMB 173005, IAM 1513, IMET 10409, JCM 2788, LMG 2089, NCIB 9393, NCTC 8545, NRRL B-1496, R. Hugh 342, USCC 1337, VKM B-893]
Species Brevundimonas diminuta
Culti-Loops are CE marked.