Learn More
Thermo Scientific™ Oxoid™ IMAGEN™ Influenza A & B
Product Code. 13255779
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13255779 50 Tests 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13255779 Supplier Thermo Scientific™ Supplier No. K6105112
  • ONLINE EXCLUSIVE PRICE
    672.00 EUR
    521.37 EUR / 50 tests
    Save 150.63 EUR
    22% Off
 Restricted Item
Estimated Shipment: 19-11-2026
For the detection and identification of Influenza virus strains A and B in clinical specimens or cell cultures
  • Test utilizes species-specific monoclonal antibodies to detect epitopes of Influenza virus glycoproteins and fusion proteins specific to either Influenza virus type A or Influenza virus type B
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 Tests