Learn More
Thermo Scientific™ Forma™ Copper Interior Kit
Description
Precisely control the CO2 and temperature with Thermo Scientific™ Forma™ Series CO2 Incubators. Forma™ CO2 Incubator Accessories are important components for optimal operation.
- Precise control
- Choice of TC or IR sensors
- Superior parameter recovery characteristics
- Continuous contamination control technology
Specifications
Specifications
| CO2 Sensor Technology | TC Sensor or IR Sensor |
| For Use With (Equipment) | Forma Steri-cycle CO2 incubator |
| Lighting | LED |
| Material | Copper |
| Type | Copper Interior Kit |