Learn More
Thermo Scientific™ Fixed Needle Autosampler Syringes, 2500uL, for Thermo Scientific™ Instruments
Product Code. 13446886 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
2.5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
13446886 2.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13446886 Supplier Thermo Scientific™ Supplier No. 365Q2131
Big Savings for Better Separation
Use promo code 27992
View all promotions
Estimated Shipment: 12-11-2026
Gas-tight, headspace.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Description 2.5 mL FN GT HS, 23 G, 65 mm, 115°C
Needle Gauge 23 G
Length (Metric) 65 mm
Product Type GC Autosampler Syringe
Max. Temperature (Metric) 115°C
Quantity Ea.
Volume (Metric) 2.5 mL
Length (Metric) Needle 65 mm
Needle Point Style Side Hole
For Use With (Equipment) TriPlus RSH Autosampler
PTFE Tipped Plunger Yes