Learn More
Thermo Scientific™ 24-Well Semi-Skirted PCR Plate
Product Code. 10563795
Change view
Click to view available options
Color:
Red
White
Yellow
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10563795 Red Pack of 50
10405353 Yellow Pack of 50
12657506 White Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10563795 Supplier Thermo Scientific™ Supplier No. AB0624/R
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
24-well semi-skirted plate for PCR and qPCR applications.
The 24-Well Semi-Skirted PCR Plate is pre-cut into 24-well segments. Its convenient format allows for lower sample numbers. The semi skirt adds rigidity and allots space for labeling or barcoding. Available in various colors and compatible with standard multichannel pipettes. Sealing is possible with adhesive films and foils and all cap strips.
The Thermo Scientific 24-Well Semi-Skirted PCR Plate is pre-cut into 24-well segments. Its convenient format allows for lower sample numbers. The semi skirt adds rigidity and allots space for labeling or barcoding. Available in various colors and compatible with standard multichannel pipettes. Sealing is possible with adhesive films and foils and all cap strips.
Highlights
• Conveniently pre-cut into 24-well segments
• Suitable for use in 0.2 mL thermal cycler blocks
• Semi-skirt adds rigidity and allows for labeling or barcoding
• 0.3 mL maximum well volume (when used with adhesive seals)
• Sealable using adhesive films and foils and all cap strips
Certifications
Certified free from human DNA, RNases, and DNases.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 4 x 6
Color Red
Material Virgin Polypropylene
No. of Wells 24