Learn More
Thermographic Measurements Thermax Encapsulated Temperature Indicator
Product Code. 17866088
Change view
Click to view available options
Max. Temperature (English):
109°F
144°F
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Max. Temperature (English) Unit Size
17866088 144°F Pack of 50
17856078 109°F Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17866088 Supplier Thermographic Measurements Supplier No. M01ETHE062D01P1
  • ONLINE EXCLUSIVE PRICE
    12.85 EUR
    10.92 EUR / Pack of 50
    Save 1.93 EUR
    15% Off
Estimated Shipment: 11-02-2027
Single temperature indicator shows when maximum temperature has been achieved
  • Immediate Response
  • Permanent Record of Highest Temperature Achieved
  • Oil, Water, and Steam Resistant
  • Inexpensive and Easy to Use
  • Self-adhesive
Specifications
Height (Metric) 13 mm
Humidity Range 40 to 60%
Length (Metric) 18 mm
Material Polyester Film
Max. Temperature (English) 144°F
Max. Temperature (Metric) 62°C
Min. Temperature (English) 36°F
Min. Temperature (Metric) 2°C
Temperature Range (English) 144°F
Temperature Range (Metric) 62°C
For Use With (Equipment) Electrical Equipment, Pumps, Braking Systems, Water Pumps, Aeronautical Equpment, Military Equipment
Subdivision Single Value
Width (Metric) 18 mm
Scale °C and °F