Learn More
Brady™ Thermal Transfer Printer Cleaning Kit
Product Code. 12851921 Shop All Brady Products
Change view
Click to view available options
Quantity:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12851921 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12851921 Supplier Brady™ Supplier No. 133251
  • ONLINE EXCLUSIVE PRICE
    74.00 EUR
    72.26 EUR / Pack of 1
    Save 1.74 EUR
    2% Off
Estimated Shipment: 17-11-2026
Prolong printer life and improve quality. The Brady™ Thermal Transfer Printer Cleaning Kit combats the dust and dirt that naturally gathers on printer heads.
  • Use to combat the dust and dirt that naturally gathers on a printhead
  • Improve print quality and lifespan with regular cleanings
Specifications
Product Type Cleaning Kit
Alternate Part Number PCK-6
Color White
For Use With (Equipment) BBP11;BBP12;BBP30;BBP31;BBP33;BBP35;BBP37;BBP72;BBP81;BBP85;i3300;i5100;i5300;i7100;i7500;PR Plus;S3100;IP Series;THT 76 mm Core Printers;S3700;A5500;Wraptor A6200;Wraptor A6500
Model PCK-6
Quantity 1