Learn More
Theophylline-Ethylenediamine Anhydrous, Mikromol™
Product Code. 18043080
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18043080 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18043080 Supplier Mikromol Supplier No. MM1050.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Theophylline-Ethylenediamine Anhydrous
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Theophylline-Ethylenediamine Anhydrous
CAS 317-34-0
Analyte or Component Names Theophylline-Ethylenediamine Anhydrous
Shipping Condition Ice pack (-20°C)
Molecular Formula 2 C7 H8 N4 O2 . C2 H8 N2
Certifications/Compliance ISO 17034
Synonym Theophylline-Ethylenediamine, Aminophylline
InChI Formula InChI=1S/2C7H8N4O2.C2H8N2/c2*1-10-5-4(8-3-9-5)6(12)11(2)7(10)13;3-1-2-4/h2*3H,1-2H3,(H,8,9);1-4H2
SMILES CN1C(=O)N(C)c2nc[nH]c2C1=O.CN3C(=O)N(C)c4nc[nH]c4C3=O.NCCN
IUPAC Name 1,3-dimethyl-7H-purine-2,6-dione;ethane-1,2-diamine
Molecular Weight (g/mol) 420.43
Quantity 250 mg
Formula Weight 420.1982 g/mol