Learn More
Theophylline-Ethylenediamine Anhydrous, LoGiCal
Product Code. 30464000
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464000 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30464000 Supplier LoGiCal Supplier No. LGCFOR1050.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Theophylline-Ethylenediamine Anhydrous
CAS 317-34-0
Analyte or Component Names Theophylline-Ethylenediamine Anhydrous
Shipping Condition Ice pack (-20°C)
Molecular Formula 2 C7 H8 N4 O2 . C2 H8 N2
Certifications/Compliance ISO 9001
Synonym Theophylline-Ethylenediamine, Aminophylline
InChI Formula InChI=1S/2C7H8N4O2.C2H8N2/c2*1-10-5-4(8-3-9-5)6(12)11(2)7(10)13;3-1-2-4/h2*3H,1-2H3,(H,8,9);1-4H2
Solution Type Neat
SMILES CN1C(=O)N(C)c2nc[nH]c2C1=O.CN3C(=O)N(C)c4nc[nH]c4C3=O.NCCN
IUPAC Name 1,3-dimethyl-7H-purine-2,6-dione;ethane-1,2-diamine
Industry Type Clinical Testing
Molecular Weight (g/mol) 420.43
Quantity 10 mg
Formula Weight 420.1982 g/mol