missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Themis Recombinant Rabbit Monoclonal Antibody (7G7)

Product Code. 17970623
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970623 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17970623 Supplier Invitrogen™ Supplier No. MA550142
  • ONLINE EXCLUSIVE PRICE
    446.00 EUR
    428.16 EUR / 100µL
    Save 17.84 EUR
    4% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Themis Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Flow, ELISA

Themis is a cytoplasmic protein that is essential for positive selection and the development of single-positive thymocytes. Highest levels of expression are found in CD4+CD8+ double-positive thymocytes, with lower expression found in mature T cells. In addition to Grb1, Themis has been found to associate with PLC gamma 1 and LAT and is quickly phosphorylated by SLP-76 following TCR signaling. There are 5 isoforms of mouse Themis that are produced as a result of alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Themis
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Monoclonal
Clone 7G7
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Themis
Gene Accession No. Q8N1K5
Gene Alias bA325O24.3; bA325O24.4; C6orf190; C6orf207; E430004N04Rik; FLJ40584; Gasp; GRB2-associated protein; Grb2-associating protein; MGC163388; Protein THEMIS; Protein thylex; RGD1560849; signaling phosphoprotein specific for T cells; SPOT; THEMIS; thylex; thymocyte expressed molecule involved in selection; thymocyte selection associated; thymocyte selection pathway associated; thymocyte-expressed molecule involved in selection; Tsepa
Gene Symbols Themis
Host Species Rabbit
Immunogen A synthesized peptide derived from Human THEMIS.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387357
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.