missing translation for 'onlineSavingsMsg'
Learn More

Thapsigargin, 99% (TLC), 97.6% (HPLC), MP Biomedicals™

Product Code. 11444282 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444282 5 mg 5mg
11434282 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444282 Supplier MP Biomedicals Supplier No. 0215899905
  • ONLINE EXCLUSIVE PRICE
    1155.00 EUR
    1098.30 EUR / 5mg
    Save 56.70 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 67526-95-8
Molecular Formula C34H50O12
Molecular Weight (g/mol) 650.76
MDL Number MFCD00083511
InChI Key IXFPJGBNCFXKPI-FSIHEZPISA-N
Synonym thapsigargin, unii-z96bq26rzd, z96bq26rzd, chembl96926, octanoic acid 3s-3alpha, 3abeta, 4alpha, 6beta, 6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester, octanoic acid, 6-acetyloxy-2,3,3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy azuleno 4,5-b furan-7-yl ester, 3s-3alpha,3abeta,4alpha,6beta,6abeta,7beta,8alpha z ,9balpha, tg1, --thapsigargin, octanoic acid 3s-3alpha, 3abeta, 4alpha, 6beta,6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester, octanoic acid 3s-3alpha, 3abeta, 4alpha,6beta, 6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester
PubChem CID 126969181
IUPAC Name (3S,3aR,4S,6S,6aR,7S,8S,9bS)-6-(acetyloxy)-4-(butanoyloxy)-3,3a-dihydroxy-3,6,9-trimethyl-8-{[(2Z)-2-methylbut-2-enoyl]oxy}-2-oxo-2H,3H,3aH,4H,5H,6H,6aH,7H,8H,9bH-azuleno[4,5-b]furan-7-yl octanoate
SMILES CCCCCCCC(=O)O[C@@H]1[C@@H](OC(=O)C(\C)=C/C)C(C)=C2[C@@H]3OC(=O)[C@@](C)(O)[C@@]3(O)[C@H](C[C@](C)(OC(C)=O)[C@@H]12)OC(=O)CCC

Specifications

Color White
Assay Percent Range 99% (TLC), 97.6% (HPLC)
Quantity 5 mg
Formula Weight 650.76
Physical Form Crystalline Powder
Chemical Name or Material Thapsigargin, For TLC and HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.