missing translation for 'onlineSavingsMsg'
Learn More

Thalidomide, MP Biomedicals™

Product Code. 11499941 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
50 g
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499941 250 mg 250mg
11489941 100 mg 100mg
11479941 50 g 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499941 Supplier MP Biomedicals Supplier No. 0215875383
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 50-35-1
Molecular Formula C13H10N2O4
Molecular Weight (g/mol) 258.23
MDL Number MFCD00153873
InChI Key UEJJHQNACJXSKW-UHFFFAOYNA-N
Synonym thalidomide, thalomid, +/--thalidomide, sedoval, 2-2,6-dioxopiperidin-3-yl isoindoline-1,3-dione, corronarobetin, psycholiquid, psychotablets, theophilcholine, algosediv
PubChem CID 5426
ChEBI CHEBI:74947
IUPAC Name 2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES O=C1N(C2CCC(=O)NC2=O)C(=O)C2=CC=CC=C12

Specifications

Color White
Quantity 250 mg
Physical Form Solid
Chemical Name or Material Thalidomide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.