missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Thalidomide 4'-oxyacetamide-alkylC2-amine
GREENER_CHOICE

Product Code. 18796096
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18796096 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18796096 Supplier Tocris Bioscience™ Supplier No. 6949/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thalidomide-alkylC2-amine

Thalidomide 4'-oxyacetamide-alkylC2-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a short alkyl linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.

This product has been recently renamed. The previous name for this product was Thalidomide - linker 10

Please contact us for SD files of our available Degrader Building Blocks.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Aminoethyl)-2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]oxy]acetamide hydrochloride
CAS 2341841-02-7
Synonym Thalidomide - linker 10
Quantity 25 mg
Molecular Formula C17H18N4O6.HCl
Formula Weight 410.81
Product Type Ligand
Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.