Learn More
TGSH, TRC
Product Code. 30135084
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135084 25 g 25g
30134179 100 g 100g
17953055 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135084 Supplier Toronto Research Chemicals Supplier No. TRCT33797025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 41481-66-7
Molecular Weight (g/mol) 330.4
Synonym Phenol, 4,4'-sulfonylbis[2-(2-propenyl)- (9CI), 4,4'-Sulfonylbis[2-(2-propen-1-yl)phenol], 2,2'-Diallyl-4,4'-sulfonyldiphenol, 3,3'-Diallyl-4,4'-dihydroxybiphenyl sulfone, 3,3'-Diallyl-4,4'-dihydroxydiphenylsulfone, 4,4'-Sulfonylbis(2-allylphenol), Bis(3-allyl-4-hydroxyphenyl) sulfone, Bis(4-hydroxy-3-allylphenyl) sulfone, TG-SA, TGSH
IUPAC Name 4-(4-hydroxy-3-prop-2-enylphenyl)sulfonyl-2-prop-2-enylphenol
SMILES Oc1ccc(cc1CC=C)S(=O)(=O)c2ccc(O)c(CC=C)c2
Specifications
Molecular Formula C18 H18 O4 S
Quantity 25 g
InChI Formula InChI=1S/C18H18O4S/c1-3-5-13-11-15(7-9-17(13)19)23(21,22)16-8-10-18(20)14(12-16)6-4-2/h3-4,7-12,19-20H,1-2,5-6H2
Formula Weight 330.0926
Percent Purity >95%
Chemical Name or Material 3,3'-Diallyl-4,4'-dihydroxydiphenylsulfone
Grade HPLC