missing translation for 'onlineSavingsMsg'
Learn More

TFCP2 Antibody, Novus Biologicals™

Product Code. 30226623 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30226623 100 μL 100µL
30227596 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30226623 Supplier Novus Biologicals Supplier No. NBP338167100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TFCP2 Polyclonal antibody specifically detects TFCP2 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TFCP2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias alpha-globin transcription factor CP2, CP2, LBP-1C, LSF1D, LSFLBP1C, SAA3 enhancer factor, SEF, TFCP2C, transcription factor CP2, Transcription factor LSF
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human TFCP2 (NP_005644.2).,, Sequence:, MAWALKLPLADEVIESGLVQDFDASLSGIGQELGAGAYSMSDVLALPIFKQEESSLPPDNENKILPFQYVLCAATSPAVKLHDETLTYLNQGQSYEIRMLDNRKLGELPEINGKLVKSIFRVVFHDRRLQYTEHQQLEGWRWNRPGDRILDIDIPMSVGIIDPRANPTQLNTVEFLWDPAKRTSVFIQVHCISTEFTMRKHGGEKGVPFRVQIDTFKENENGEYTEHLHSASCQIKVFKPKGADRKQKTDREKMEKRTPHEKEKYQPSYETTILTECSPW
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7024
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.