Learn More
Texwipe™ TexWrite™ Clipboard
Product Code. 15722416 Shop All Texwipe Products
Change view
Click to view available options
Material:
Polypropylene, Plastic
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15722416 Polypropylene, Plastic Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15722416 Supplier Texwipe™ Supplier No. TX5835
  • ONLINE EXCLUSIVE PRICE
    614.00 EUR
    521.20 EUR / Pack of 12
    Save 92.80 EUR
    15% Off
 Restricted Item
Estimated Shipment: 04-03-2027
Ideal portable writing surface for use in critical environments
  • Corrosion- and chemical-resistant, durable polypropylene construction
  • No sharp edges to damage gloves or garments
  • Rigid back allows use while standing
  • Suitable for use with 8 1/2 × 11 in. paper
  • Securely holds up to 50 sheets of cleanroom paper
  • Cleanroom packaged
Specifications
Material Polypropylene, Plastic
For Use With (Application) 8.5 x 11 in. or A4 paper
Product Type Cleanroom Clipboard