Learn More
Texwipe™ Alpha™ 10 Sealed-Border Wipers
Product Code. 15604987 Shop All Texwipe Products
Change view
Click to view available options
Dimensions (L x W):
12 x 12 in.
9 x 9 in. (22.86 x 22.86 cm)
Product Type:
Dry Wipe
Pre-wetted Wipe
Saturation Solution:
70% IPA, 30% WFI
70% Isopropyl Alcohol
Dry Wiper
Unit Size:
Pack of 100
Pack of 250
Pack of 400
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Product Type Saturation Solution unitSize
15604987 12 x 12 in. Pre-wetted Wipe 70% IPA, 30% WFI Pack of 250
11585355 12 x 12 in. Dry Wipe Dry Wiper Pack of 100
15614987 9 x 9 in. (22.86 x 22.86 cm) Pre-wetted Wipe 70% Isopropyl Alcohol Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15604987 Supplier Texwipe™ Supplier No. TX3280
  • ONLINE EXCLUSIVE PRICE
    882.00 EUR
    752.16 EUR / Pack of 250
    Save 129.84 EUR
    14% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
For ISO Class 3 to 4 applications
  • Vectra™ processed—hands-free, continuous manufacturing purification and inspection
  • Double-knit, no-run interlock construction of 100% continuous-filament polyester
  • Patented sealed-border wiper for lowest releasable fibers and particles
  • Optimum balance of wiping efficiency, abrasion resistance, and sorbency
  • Ideal wiper for use in multiple environments
  • Lot-to-lot traceability
Order Info
Available dry or sterile and presaturated with isopropanol. Basis weight: 130g/m2. Sterile wipers 19-130-2061 packed 50 wipers/ flatpack, 5 flatpacks /case; 19-165-653 packed 20 wipers per flexpack, 20 flexpacks/case.
Specifications
Product Type Pre-wetted Wipe
Length (English) 12 in.
Length (Metric) 30.5 cm
Width (English) 12 in.
Width (Metric) 30.5 cm
Material Polyester
Certifications/Compliance ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
Edges Sealed
Dimensions (L x W) 12 x 12 in.
For Use With (Application) Ideal wipes for use in multiple environments
Saturation Solution 70% IPA, 30% WFI