missing translation for 'onlineSavingsMsg'
Learn More

Tetraxetan, Mikromol™

Product Code. 17901176
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17901176 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17901176 Supplier Mikromol Supplier No. MM3916.000250
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Tetraxetan
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DOTA
CAS 60239-18-1
Analyte or Component Names DOTA
Shipping Condition Room Temperature
Molecular Formula C16 H28 N4 O8
Certifications/Compliance ISO 17025
Synonym 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid, 1,4,7,10-Tetra(carboxymethyl)-1,4,7,10-tetraazacyclododecane, 1,4,7,10-Tetraazacyclodecane-1,4,7,10-tetraacetic acid, 1,4,7,10-Tetraazacyclododecane-N,N',N',N''-tetraacetic acid, DOTA, NSC 681107, Tetraxetan
InChI Formula InChI=1S/C16H28N4O8/c21-13(22)9-17-1-2-18(10-14(23)24)5-6-20(12-16(27)28)8-7-19(4-3-17)11-15(25)26/h1-12H2,(H,21,22)(H,23,24)(H,25,26)(H,27,28)
SMILES OC(=O)CN1CCN(CC(=O)O)CCN(CC(=O)O)CCN(CC(=O)O)CC1
IUPAC Name 2-[4,7,10-tris(carboxymethyl)-1,4,7,10-tetrazacyclododec-1-yl]acetic acid
Molecular Weight (g/mol) 404.42
Quantity 250 mg
Formula Weight 404.1907 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.