Learn More
Tetramethylammonium Borohydride, TRC
Specifications
Specifications
| Chemical Name or Material | Tetramethylammonium Borohydride |
| CAS | 16883-45-7 |
| Molecular Formula | C4 H12 N . B |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C4H12N.B/c1-5(2,3)4;/h1-4H3;/q+1;-1 |
| SMILES | [B-].C[N+](C)(C)C |
| IUPAC Name | boron(1-);tetramethylazanium |
| Molecular Weight (g/mol) | 84.956 |
| Formula Weight | 85.106 |