Learn More
Tetramethrin, TRC
Product Code. 30409317
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409317 1 g -
30415105 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30409317 Supplier Toronto Research Chemicals Supplier No. TRCT2986901G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7696-12-0
Molecular Formula C19 H25 N O4
Molecular Weight (g/mol) 331.41
Synonym Tetramethrin, Phthalthrin, Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propen-1-yl)-, (1,3,4,5,6,7-hexahydro-1,3-dioxo-2H-isoindol-2-yl)methyl ester (ACI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methyl-1-propenyl)-, (1,3,4,5,6,7-hexahydro-1,3-dioxo-2H-isoindol-2-yl)methyl ester (9CI), Cyclopropanecarboxylic acid, 2,2-dimethyl-3-(2-methylpropenyl)-, ester with N-(hydroxymethyl)-1-cyclohexene-1,2-dicarboximide (7CI, 8CI), (1,3-Dioxo-4,5,6,7-tetrahydro-1H-isoindol-2(3H)-yl)methyl 2,2-dimethyl-3-(2-methylprop-1-en-1-yl)cyclopropanecarboxylate, (1-Cyclohexene-1,2-dicarboximido)methyl chrysanthemate, (1-Cyclohexene-1,2-dicarboximido)methyl chrysanthemumate, Bioneo-Pynamin, d-Tetramethrin, Insectol, Neo-Pynamin, Neo-Pynamin Forte, Phthalthrin, Pibutox, Pion 1, Pyrethrol L, Tetramethrin
IUPAC Name (1,3-dioxo-4,5,6,7-tetrahydroisoindol-2-yl)methyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
SMILES CC(=CC1C(C(=O)OCN2C(=O)C3=C(CCCC3)C2=O)C1(C)C)C
Tekniske data
Quantity 1 g
InChI Formula InChI=1S/C19H25NO4/c1-11(2)9-14-15(19(14,3)4)18(23)24-10-20-16(21)12-7-5-6-8-13(12)17(20)22/h9,14-15H,5-8,10H2,1-4H3
Formula Weight 331.1784
Chemical Name or Material Tetramethrin
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.