missing translation for 'onlineSavingsMsg'
Learn More

alpha-Tetralone, 98%

Product Code. p-197900 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10011620 25 g Glass bottle 25g
10284800 100 g Glass bottle 100g
10337070 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10011620 Supplier Thermo Scientific Acros Supplier No. 138310250
Add to basket
This item is not returnable. View return policy

CAS: 529-34-0 | C10H10O | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-34-0
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001688
InChI Key XHLHPRDBBAGVEG-UHFFFAOYSA-N
Synonym alpha-tetralone, 1-tetralone, 3,4-dihydronaphthalen-1 2h-one, 1,2,3,4-tetrahydronaphthalen-1-one, 3,4-dihydro-1 2h-naphthalenone, tetralone, a-tetralone, 1-oxotetralin, 1 2h-naphthalenone, 3,4-dihydro, .alpha.-tetralone
PubChem CID 10724
IUPAC Name 3,4-dihydro-2H-naphthalen-1-one
SMILES O=C1CCCC2=CC=CC=C12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 5.0°C to 6.0°C
Color Brown to Yellow
Density 1.0960g/mL
Boiling Point 127.0°C (13.0 mmHg)
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5675 to 1.5695
Quantity 25 g
Beilstein 07, 370
Specific Gravity 1.096
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hexane
Formula Weight 146.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material α-Tetralone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.