missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(dimethylamino)silane, 99%

Product Code. 11496160
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11496160 10 g 10g
11486160 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496160 Supplier Thermo Scientific Alfa Aesar Supplier No. H27092.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1624-01-7 | C8H24N4Si | 204.39 g/mol

Silicon dioxide thin films prepared by chemical vapor deposition from tetrakis (dimethylamino) silane and ozone. Aminosilanes mere prepared by methods all essentially similar, and only the 15olation of tetrakis-(dimethylamino)-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon dioxide thin films prepared by chemical vapor deposition from tetrakis (dimethylamino) silane and ozone. Aminosilanes mere prepared by methods all essentially similar, and only the 15olation of tetrakis-(dimethylamino)-silane.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1624-01-7
Molecular Formula C8H24N4Si
Molecular Weight (g/mol) 204.39
MDL Number MFCD00014859
InChI Key SSCVMVQLICADPI-UHFFFAOYSA-N
Synonym tetrakis dimethylamino silane, silanetetramine, octamethyl, octamethylsilanetetramine, octamethylsilatetramine, silane, tetrakis dimethylamino, silane 48-12 tetrakis, dimethylamine, n,n',n,n'-silanetetrayltetrakis, silanetetramine, n,n,n',n',n,n,n',n'-octamethyl, n,n,n',n',n,n,n',n'-octamethylsilanetetraamine, si nme2 4
PubChem CID 74194
IUPAC Name N-methyl-N-[tris(dimethylamino)silyl]methanamine
SMILES CN(C)[Si](N(C)C)(N(C)C)N(C)C

Specifications

Boiling Point 196°C
Melting Point 15°C
Quantity 10 g
Linear Formula [(CH3)2N]4Si
UN Number UN3129
Flash Point 58°C (136°F)
Solubility Information Reacts with water.
Formula Weight 204.4
Percent Purity 99%
Odor Amine-like
Refractive Index 1.443
Sensitivity Moisture sensitive
Density 0.885
Chemical Name or Material Tetrakis(dimethylamino)silane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.