missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, TRC

Product Code. 17981375
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17981375 500 mL 500mL
17934755 1 L 1 L
17973815 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17981375 Supplier Toronto Research Chemicals Supplier No. TRCT293518500ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4 H8 O
Molecular Weight (g/mol) 72.11
Synonym Tetrahydrofuran, Furan, tetrahydro-, Tetrahydrofuran, 2,3,4,5-Tetrahydrofuran, Butane α,δ-oxide, Butane, 1,4-epoxy-, Cyclotetramethylene oxide, Furanidine, NSC 57858, Oxacyclopentane, Oxolane, THF, Tetramethylene oxide
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Quantity 500 mL
InChI Formula InChI=1S/C4H8O/c1-2-4-5-3-1/h1-4H2
Formula Weight 72.0575
Chemical Name or Material Tetrahydrofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.