Learn More
Tetrahydrobiopterin, MedChemExpress
Product Code. 16476817
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476817 5 mg 5mg
16474118 10 mg 10mg
16460477 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16476817 Supplier Medchem Express Supplier No. HY107383
MedChemExpress Tetrahydrobiopterin ((Rac)-Sapropterin) is a cofactor of the aromatic amino acid hydroxylases enzymes and also acts as an essential cofactor for all nitric oxide synthase (NOS) isoforms.
Tetrahydrobiopterin ((Rac)-Sapropterin) is a cofactor of the aromatic amino acid hydroxylases enzymes and also acts as an essential cofactor for all nitric oxide synthase (NOS) isoforms.
Specifications
Chemical Name or Material Tetrahydrobiopterin
Physical Form Solid
CAS 17528-72-2
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C9H15N5O3
For Use With (Application) COVID-19-immunoregulation
Synonym (Rac)-Sapropterin
Solubility Information DMSO : 50 mg/mL (207.25 mM; Need ultrasonic)
SMILES O=C1C2=C(NC(N)=N1)NCC(C(O)C(O)C)N2
Molecular Weight (g/mol) 241.25
Formula Weight 241.25
Percent Purity 99.72%
Grade Research