missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2H-pyran-2-ol, >90%, TRC

Product Code. 30450795
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450795 10 mg -
30452321 100 mg 100mg
30450772 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450795 Supplier Toronto Research Chemicals Supplier No. TRCT89020510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-54-2
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
Synonym Tetrahydro-2H-pyran-2-ol
IUPAC Name oxan-2-ol
SMILES OC1CCCCO1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C5H10O2/c6-5-3-1-2-4-7-5/h5-6H,1-4H2
Formula Weight 102.0681
Chemical Name or Material Tetrahydro-2H-pyran-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.