Learn More
Tetraethylammonium Tetrafluoroborate, Sigma-Aldrich™
Specifications
Specifications
| CAS | 429-06-1 |
| Melting Point | 365°C to 368°C |
| Molecular Formula | (C2H5)4NBF4 |
| MDL Number | MFCD00011827 |
| Quantity | 25 g |
| Synonym | Tetraethylammonium tetrafluoroborate, Tetraethylammonium tetrafluoroborate |
| Molecular Weight (g/mol) | 217.06 |
| Grade | Research |