Learn More
Tetraethylammonium Bromide, Sigma-Aldrich™
Description
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This advanced-grade supporting electrolyte is used for electrochemical applications, enabling efficient charge transfer and enhanced performance, offering a greener, potentially more sustainable option for specialized electrochemical processes.
Specifications
Specifications
| Melting Point | >285°C |
| Density | 1.397 g/cm3 at 20°C |
| Molecular Formula | (C2H5)4NBr |
| MDL Number | MFCD00011825 |
| Quantity | 1 kg |
| Synonym | Tetraethylammonium bromide, Tetraethylammonium bromide |
| Molecular Weight (g/mol) | 210.16 |
| Grade | Research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.