missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, Supelco™

Product Code. 15896910
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
190 L
Unit Size:
1 L
190 L
2.5 L
25 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896910 25 L 25 L
15863980 1 L 1 L
15873980 2.5 L 2.5 L
15806920 190 L 190 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15896910 Supplier Supelco Supplier No. 1009646025
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EMPLURA™

EMPLURA™ is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-18-4

Specifications

Melting Point -22°C
Density 1.62 g/cm3 at 20°C
Boiling Point 121°C/1013 hPa
MDL Number MFCD00000834
Quantity 25 L
Synonym Tetrachloroethylene, Ethylene tetrachloride, Perchloroethylene, PCE, Tetrachloroethylene
Molecular Weight (g/mol) 165.83
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.