Learn More
Tetrachloroethylene, Supelco™
Description
Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified.
Specifications
Specifications
| Melting Point | -22°C |
| Density | 1.62 g/cm3 at 20°C |
| Boiling Point | 121°C/1013 hPa |
| MDL Number | MFCD00000834 |
| Quantity | 2.5 L |
| Synonym | Tetrachloroethylene, Ethylene tetrachloride, Perchloroethylene, PCE, Tetrachloroethylene |
| Molecular Weight (g/mol) | 165.83 |
| Grade | Research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.