missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Phosphate, ∽97%, MP Biomedicals™

Product Code. 11455472 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455472 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11455472 Supplier MP Biomedicals Supplier No. 0219551301
  • ONLINE EXCLUSIVE PRICE
    71.65 EUR
    68.25 EUR / 1g
    Save 3.40 EUR
    4% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ion-associating reagent for HPLC

Specifications

CAS 5574-97-0
Color White
Assay Percent Range Approx. 97%
Molecular Formula C16H38NO4P
MDL Number MFCD00064526
Quantity 1 g
Synonym tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
InChI Key ARRNBPCNZJXHRJ-UHFFFAOYSA-M
SMILES OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium dihydrogen phosphate
Molecular Weight (g/mol) 339.46
PubChem CID 2735142
Formula Weight 339.46
Physical Form Powder
Chemical Name or Material Tetrabutylammonium phosphate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.