missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium phosphate, ∽97%, MP Biomedicals™

Product Code. 11465472 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11465472 5 g 5g
11475472 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465472 Supplier MP Biomedicals Supplier No. 0219551305
  • ONLINE EXCLUSIVE PRICE
    187.00 EUR
    177.45 EUR / 5g
    Save 9.55 EUR
    5% Off
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 5574-97-0
Molecular Formula C16H38NO4P
Molecular Weight (g/mol) 339.46
MDL Number MFCD00064526
InChI Key ARRNBPCNZJXHRJ-UHFFFAOYSA-M
Synonym tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
PubChem CID 2735142
IUPAC Name tetrabutylazanium dihydrogen phosphate
SMILES OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Color White
Quantity 5 g
Formula Weight 339.5
Percent Purity ∼97%
Physical Form Waxy Solid
Chemical Name or Material Tetrabutylammonium phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.