Learn More
Tetrabutylammonium Dibromochloride, TRC
Product Code. 30457066
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457066 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30457066 Supplier Toronto Research Chemicals Supplier No. TRCT3107832.5G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tetrabutylammonium Dibromochloride
CAS 64531-21-1
Molecular Formula C16H36Br2ClN
Quantity 2.5 g
Synonym 1-Butanaminium','N,N,N-tributyl-','bromochlorobromate(1-) (9CI), Bromate(1-)','bromochloro-','N,N,N-tributyl-1-butanaminium (9CI), N,N,N,N-Tetrabutylammonium dibromochloride, Tetrabutylammonium dibromochloride
InChI Formula InChI=1S/C16H36N.Br2Cl/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-2-3/h5-16H2,1-4H3;/q+1;-1
SMILES CCCC[N+](CCCC)(CCCC)CCCC.Br[Br-]Cl
Molecular Weight (g/mol) 437.72
Formula Weight 435.0903