missing translation for 'onlineSavingsMsg'
Learn More

Tetrabenazine-d6, TRC

Product Code. 30421998
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421998 1 mg -
30409757 2.5 mg 2.5mg
30419812 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30421998 Supplier Toronto Research Chemicals Supplier No. TRCT2840021MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1392826-25-3
Molecular Formula C19H21D6NO3
Molecular Weight (g/mol) 323.46
Synonym (3R,11bR)-rel-1,3,4,6,7,11b-Hexahydro-9,10-dimethoxy-3-(2-methylpropyl)-2H-benzo[a]quinolizin-2-one-d6, cis-2-Oxo-3-(isobutyl)-9,10-dimethoxy-1,2,3,4,6,7-hexahydro -11bH-benzo[a]quinolizine-d6, (+/-)-Tetrabenazine-d6, NSC 169886-d6, NSC 172187-d6, Ro 1-9569-d6, Rubigen-d6
SMILES O=C1C(CC(C)C)CN2CCC3=CC(OC([2H])([2H])[2H])=C(OC([2H])([2H])[2H])C=C3C2C1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C19H27NO3/c1-12(2)7-14-11-20-6-5-13-8-18(22-3)19(23-4)9-15(13)16(20)10-17(14)21/h8-9,12,14,16H,5-7,10-11H2,1-4H3/t14-,16-/m1/s1/i3D3,4D3
Formula Weight 323.2368
Chemical Name or Material Tetrabenazine-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.