Learn More
(-)-Tetrabenazine (>75% ee), TRC
Product Code. 30421690
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30421690 25 mg
30414328 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30421690 Supplier Toronto Research Chemicals Supplier No. TRCT28402325MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1026016-84-1
Molecular Formula C19H27NO3
Molecular Weight (g/mol) 317.42
Synonym (3S,11bS)-1,3,4,6,7,11b-Hexahydro-9,10-dimethoxy-3-(2-methylpropyl)-2H-benzo[a]quinolizin-2-one, (-)-TBZ, (-)-Tetrabenazine, (3S,11bS)-TBZ, (3S,11bS)-Tetrabenazine
IUPAC Name (3S,11bS)-9,10-dimethoxy-3-(2-methylpropyl)-1,3,4,6,7,11b-hexahydrobenzo[a]quinolizin-2-one
SMILES COc1cc2CCN3C[C@H](CC(C)C)C(=O)C[C@H]3c2cc1OC
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C19H27NO3/c1-12(2)7-14-11-20-6-5-13-8-18(22-3)19(23-4)9-15(13)16(20)10-17(14)21/h8-9,12,14,16H,5-7,10-11H2,1-4H3/t14-,16-/m0/s1
Formula Weight 317.1991
Chemical Name or Material (-)-Tetrabenazine