Learn More
Tetra-n-butylammonium Hydrogen Sulfate, Sigma-Aldrich™
Product Code. 15816620
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15816620 500 g 500g
15806620 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15816620 Supplier Sigma Aldrich Supplier No. 8188580500
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32503-27-8
Specifications
Melting Point 168°C to 174°C
MDL Number MFCD00011637
Quantity 500 g
Synonym Tetra-n-butylammonium hydrogen sulfate, Tetra-n-butylammonium hydrogen sulfate
Molecular Weight (g/mol) 339.53
Grade Research