missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium hydrogen sulfate, LiChropur™, EMD-Millipore

Product Code. 15895890
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15895890 5 g Glass 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15895890 Supplier Supelco Supplier No. 1183120025
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 32503-27-8
Density 650kg/m3
Packaging Glass
Molecular Formula C16H37NO4S
Quantity 5 g
Synonym tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
InChI Key SHFJWMWCIHQNCP-UHFFFAOYSA-M
SMILES OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium hydrogen sulfate
Molecular Weight (g/mol) 339.54
PubChem CID 94433
Formula Weight 339.54
Chemical Name or Material LiChropur Tetra-n-butylammonium hydrogen sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.