missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium Fluoride Trihydrate, Sigma-Aldrich™

Product Code. 15870281
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15870281 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15870281 Supplier Sigma Aldrich Supplier No. 8181640010
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 87749-50-6
Melting Point 62°C to 63°C
Molecular Formula (CH3CH2CH2CH2)4NF · 3H2O
MDL Number MFCD00149981
Quantity 10 g
Synonym Tetra-n-butylammonium fluoride trihydrate, TBAF, Tetra-n-butylammonium fluoride trihydrate
Molecular Weight (g/mol) 315.51
Grade Research
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.