missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium Chloride, Sigma-Aldrich™

Product Code. 15830051
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15830051 100 g 100g
15820051 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15830051 Supplier Sigma Aldrich Supplier No. 8146450100
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Find details here.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1112-67-0

Specifications

Melting Point 41°C to 44°C
MDL Number MFCD00011635
Quantity 100 g
Synonym Tetra-n-butylammonium chloride, Tetra-n-butylammonium chloride
Molecular Weight (g/mol) 277.92
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.