Learn More
Velp Scientifica™ Test Tubes for Digestion Units
Product Code. 11725654 Shop All Velp Products
Change view
Click to view available options
For Use With (Equipment):
DK6/48 and UDK series
Digesters DK 20/26, DKL 42/26 and UDK Distillation Units
UDK Series distillation units
Diameter (Metric) Outer:
26 mm
42 mm
48 mm
80 mm
Unit Size:
Each
Pack of 3
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Diameter (Metric) Outer Unit Size
11725654 Digesters DK 20/26, DKL 42/26 and UDK Distillation Units 26 mm Pack of 6
11900114 DK6/48 and UDK series 48 mm Each
10777925 UDK Series distillation units 80 mm Each
11715654 - 42 mm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11725654 Supplier Velp Scientifica™ Supplier No. A00000146
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
Estimated Shipment: 23-11-2026
Includes
6 Test Tubes/Box
Facilitates the nitrogen analysis and protein determination process. Velp Scientifica™ Test Tubes for Digestion Units are for use with Velp Scientifica™ Kjeldahl Digestion Units.
  • Available in different counts
  • Available in different sizes and diameters
Specifications
Product Type Test Tubes
Capacity (Metric) 100 mL
Diameter (Metric) Outer 26 mm
For Use With (Equipment) Digesters DK 20/26, DKL 42/26 and UDK Distillation Units
Quantity 6 Tubes/ Box