Learn More
Fisherbrand™ Foam Stands for Vortex Mixer
Product Code. 12365107 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
16 mm Tubes
19 Microtubes
29 mm Tubes
No. of Positions:
4
5
19
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds No. of Positions Unit Size
12365107 19 Microtubes 19 Each
11840992 29 mm Tubes 4 Each
11712583 16 mm Tubes 5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12365107
  • ONLINE EXCLUSIVE PRICE
    35.55 EUR
    34.20 EUR / Pack of 1
    Save 1.35 EUR
    3% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
Securely holds test tubes and vials. Velp Scientifica™ Foam Stands are compatible with Velp Scientifica Vortex Mixers.
  • Soft foam to prevent breakage
  • Suitable for different mixing requirements
Specifications
Holds 19 Microtubes
Product Type Foam Tube Rack
Color Black
For Use With (Equipment) CLASSIC, ZX3, WIZARD, ZX4, TX4
No. of Positions 19