Learn More
Fisherbrand™ Foam Stands for Vortex Mixer
Product Code. 11840992 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Holds:
16 mm Tubes
19 Microtubes
29 mm Tubes
No. of Positions:
4
5
19
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds No. of Positions Unit Size
11840992 29 mm Tubes 4 Each
11712583 16 mm Tubes 5 Each
12365107 19 Microtubes 19 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11840992
  • ONLINE EXCLUSIVE PRICE
    39.35 EUR
    38.73 EUR / Pack of 1
    Save 0.62 EUR
    1% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
Securely holds test tubes and vials. Velp Scientifica™ Foam Stands are compatible with Velp Scientifica Vortex Mixers.
  • Black in color
  • Soft foam to prevent breakage
  • Available in different diameters and capacities
Specifications
Holds 29 mm Tubes
Tube Size 29 mm
Product Type Foam Tube Rack
For Use With (Equipment) CLASSIC, ZX3, WIZARD, ZX4, TX4
No. of Positions 4