Learn More
Kartell SpA Test Tube Caps
Product Code. 11381195
Change view
Click to view available options
Color:
Blue
Green
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11381195 Blue Pack of 1000
11311205 Green Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11381195 Supplier Kartell SpA Supplier No. 8830904
  • ONLINE EXCLUSIVE PRICE
    18.55 EUR
    17.14 EUR / Pack of 1000
    Save 1.41 EUR
    7% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Finned plug for disposable test tubes
Carefully designed finned plugs for disposable tubes that are 11 to 13 mm in diameter.
Specifications
Closure Size 11 to 13 mm
Product Type Finned Plug
Autoclavable Not Autoclavable
Material LDPE
Color Blue
Sterility Non-sterile
Disposable Single-use
Fit for Purpose Certified Quality