Learn More
Reitenspiess™ Test Tube Brush
Product Code. 11337494
Click to view available options
Length (Metric):
100 mm
190 mm
230 mm
Unit Size:
Each
Diameter (Metric) Bristle:
16 mm
26 mm
Product Code. 11337494
Brand: Reitenspiess™ 10/7
Only null left
Add to basket
Edge
Add to basket
Use a Reitenspiess™ Test Tube Brush for cleaning, disinfecting and sterilizing test tubes as necessary in the laboratory. .
  • Natural bristle
  • Various lengths available
  • Various diameters
Specifications
Length (Metric) 100 mm
Quantity 1 Ea.