Learn More
Retsch™ Test Sieve Lid
Product Code. 15413223 Shop All Retsch Products
Change view
Click to view available options
For Use With (Equipment):
Test Sieve
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15413223 Test Sieve Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15413223 Supplier Retsch™ Supplier No. 69.540.0050
  • ONLINE EXCLUSIVE PRICE
    523.00 EUR
    152.01 EUR / Pack of 1
    Save 370.99 EUR
    70% Off
Estimated Shipment: 08-04-2027
Retsch™ Test Sieve Lid is constructed of stainless steel for durability.
  • Stainless steel
  • 400 mm diameter
  • For use with test sieves
Specifications
Product Type Lid
Material Stainless Steel
Diameter (Metric) 400 mm
For Use With (Equipment) Test Sieve