Learn More
tert-Butyl ((S)-4-Methyl-1-((R)-2-methyloxiran-2-yl)-1-oxopentan-2-yl)carbamate, TRC
Specifications
Specifications
| Chemical Name or Material | tert-Butyl ((S)-4-Methyl-1-((R)-2-methyloxiran-2-yl)-1-oxopentan-2-yl)carbamate |
| CAS | 247068-82-2 |
| Molecular Formula | C14H25NO4 |
| Quantity | 50 mg |
| Synonym | N-[(1S)-3-Methyl-1-[[(2R)-2-methyl-2-oxiranyl]carbonyl]butyl]carbamic Acid 1,1-Dimethylethyl Ester, [(1S)-3-Methyl-1-[[(2R)-2-methyloxiranyl]carbonyl]butyl]carbamic Acid 1,1-Dimethylethyl Ester, (2R)-2-[(2S)-2-(tert-Butoxycarbonylamino)-4-methylpentanoyl]-2-methyloxirane |
| InChI Formula | InChI=1S/C14H25NO4/c1-9(2)7-10(11(16)14(6)8-18-14)15-12(17)19-13(3,4)5/h9-10H,7-8H2,1-6H3,(H,15,17)/t10-,14+/m0/s1 |
| SMILES | CC(C)CC(NC(=O)OC(C)(C)C)C(=O)[C@@]1(C)CO1 |
| IUPAC Name | tert-butyl N-[(2S)-4-methyl-1-[(2R)-2-methyloxiran-2-yl]-1-oxopentan-2-yl]carbamate |
| Molecular Weight (g/mol) | 271.35 |